Learn More
Blooms Syndrome Protein Blm (p Ser1296) Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244115
Catalog No. NBP244115
Add to cart
Description
Description
Specifically detects Blooms Syndrome Protein Blm (p Ser1296) in Human samples, and it is validated for ELISA, Western Blotting
Blooms Syndrome Protein Blm (p Ser1296) Polyclonal antibody specifically detects Blooms Syndrome Protein Blm (p Ser1296) in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Blooms Syndrome Protein Blm (p Ser1296)
LYOPH
Western Blot 1ug/ml, ELISA 1: 10000
Affinity Purified
BLM
Anti-BLM [p Ser1290] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the internal region of human Bloom syndrome protein.
Immunogen affinity purified
RUO
Store lyophilized antibody at -20°C prior to opening. Aliquot reconstituted liquid and and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing.
Polyclonal
Reconstitute with 100 ul deionized water.
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with No Preservative
Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2
Rabbit
IgG
0.1 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary
641
Human, Mouse