Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Blooms Syndrome Protein Blm (p Thr122), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244118

Catalog No. NBP244118


Add to cart

Description

Description

Specifically detects Blooms Syndrome Protein Blm (p Thr122) in Human samples, and it is validated for ELISA, Western Blotting

Blooms Syndrome Protein Blm p Thr122 Polyclonal antibody specifically detects Blooms Syndrome Protein Blm p Thr122 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Blooms Syndrome Protein Blm (p Thr122)
1 mg/ml
Western Blot 1ug/ml, ELISA 1: 10000
Affinity Purified
BLM
Anti-BLM [p Thr122] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the internal region of human Bloom syndrome protein.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative
Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2
Rabbit
IgG
0.05 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary
641
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only