Learn More
BMP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against BMP2.
Manufacturer:  Abnova Corporation PAB16170
Catalog No. 89-118-063
Add to cart
Description
Description
The protein encoded by this gene belongs to the transforming growth factor-beta (TGFB) superfamily. The encoded protein acts as a disulfide-linked homodimer and induces bone and cartilage formation. [provided by RefSeq
Specifications
Specifications
BMP2
Unconjugated
ELISA (1-2 ug/mL) Western Blot (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user.
BMP2
Liquid
BMP2
Human BMP2.
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human, Murine, Rat
ELISA, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against BMP2.
Lyophilized from PBS.
P12643
BMP2A
Rabbit
IgG
RUO
Primary
650