Learn More
BMX Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BMX.
Supplier: Abnova Corporation PAB27109
Description
This gene encodes a nonreceptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified
Specifications
BMX | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BMX. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
P51813 | |
BMX | |
A synthetic peptide corresponding to human BMX. | |
100 μL | |
Primary | |
BMX polyclonal antibody detects endogenous levels of BMX protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
BMX | |
ETK/PSCTK2/PSCTK3 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
660 | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only