Learn More
BNIP3 Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.
Supplier: Abnova Corporation PAB7995
Description
This gene is a member of the BCL2/adenovirus E1B 19kDa-interacting protein (BNIP) family. It interacts with the E1B 19kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2. [provided by RefSeq
Specifications
BNIP3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BNIP3. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
NIP3 | |
Rabbit | |
Affinity purification | |
RUO | |
664 | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) Immunocytochemistry (0.5-1 ug/mL) The optimal working dilution should be determine | |
BNIP3 | |
BNIP3 | |
A synthetic peptide corresponding to amino acids at C-terminus of human BNIP3. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only