Learn More
BRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BRS3.
Manufacturer:  Abnova Corporation PAB16270
Catalog No. 89-118-145
Add to cart
Description
Description
Mammalian bombesin-like peptides (see MIM 137260) are widely distributed in the central nervous system as well as in the gastrointestinal tract, where they modulate smooth-muscle contraction, exocrine and endocrine processes, metabolism, and behavior. They bind to G protein-coupled receptors on the cell surface to elicit their effects. Bombesin-like peptide receptors include gastrin-releasing peptide receptor (MIM 305670), neuromedin B receptor (MIM 162341), and bombesin-like receptor-3 (BRS3) (Ohki-Hamazaki et al., 1997).[supplied by OMIM
Specifications
Specifications
BRS3
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
BRS3
Liquid
Rabbit
Immunoaffinity purification
RUO
Primary
680
Human, Murine, Rat
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of BRS3.
In PBS (0.09% sodium azide)
P32247
BRS3
A synthetic peptide (conjugated with KLH) corresponding to human BRS3.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
C-terminal Cytoplasmic domain of human.