Learn More
BST2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BST2.
Manufacturer:  Abnova Corporation PAB13047
Catalog No. 89-116-299
Add to cart
Description
Description
Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis. [provided by RefSeq
Specifications
Specifications
BST2
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
BST2
Liquid
BST2
A synthetic peptide corresponding to C-terminus 16 amino acids of human BST2.
RUO
Primary
684
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of BST2.
In PBS (0.02% sodium azide)
AAH87949
CD317
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine