Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc tag Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138046 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-046 Supplier Abnova Corporation Supplier No. PAB15370

Rabbit polyclonal antibody raised against synthetic peptides of c-Myc tag.

Sequence: EQKLISEEDL

Specifications

Antigen c-Myc tag
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to c-Myc tag.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity By ELISA, This antibody reacts with the peptide conjugated to BSA (EQKLISEEDL-BSA).
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.