Learn More
C9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of C9.
Manufacturer:  Abnova Corporation PAB25201
Catalog No. 89-126-872
Add to cart
Description
Description
This gene encodes the final component of the complement system. It participates in the formation of the Membrane Attack Complex (MAC). The MAC assembles on bacterial membranes to form a pore, permitting disruption of bacterial membrane organization. Mutations in this gene cause component C9 deficiency. [provided by RefSeq
Specifications
Specifications
C9
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of C9.
In PBS, pH 7.2 (0.05% sodium azide)
Liquid
Rabbit
Affinity purification
RUO
Primary
735
Human
Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
C9
C9
A synthetic peptide corresponding to C9.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
C9 polyclonal antibody detects endogenous levels of C9 protein.