Learn More
CA9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CA9.
Supplier:  Abnova Corporation PAB4679
Catalog No. 89-131-520
Add to Cart
Description
Description
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA IX is a transmembrane protein and the only tumor-associated carbonic anhydrase isoenzyme known. It is expressed in all clear-cell renal cell carcinoma, but is not detected in normal kidney or most other normal tissues. It may be involved in cell proliferation and transformation. This gene was mapped to 17q21.2 by fluorescence in situ hybridization, however, radiation hybrid mapping localized it to 9p13-p12. [provided by RefSeq
Specifications
Specifications
CA9
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of CA9.
In PBS (0.09% sodium azide)
NP_001207;Q16790
CA9
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CA9.
400 μL
Primary
Human
Liquid
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
CA9
CAIX/MN
Rabbit
Protein G purification
RUO
768
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions