Learn More
CAD (phospho T456), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAD.
Supplier:  Abnova Corporation PAB29236
Catalog No. 89-130-671
Add to Cart
Description
Description
The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides. [provided by RefSeq
Specifications
Specifications
CAD
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CAD.
In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
P27708
Rabbit
Affinity purification
RUO
790
Human, Mouse
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
CAD
CAD
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T456 of human CAD.
100 μL
Primary
This antibody detects endogenous levels of CAD only when phosphorylated at threonine 456.
Store at -20°C.
Avoid repeated freezing and thawing.
Recommendations
Recommendations