Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALB2 Rabbit anti-Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant CALB2.

Manufacturer:  Abnova Corporation PAB13469

Catalog No. 89-116-694


Add to cart

Description

Description

This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily. Members of this protein family have six EF-hand domains which bind calcium. Three alternatively spliced transcript variants that encode different proteins have been described. [provided by RefSeq

Specifications

Specifications

CALB2
Unconjugated
Immunohistochemistry (1:40-1:50) The optimal working dilution should be determined by the end user.
CALB2
CAL2
Rabbit
1 mL
Store at 4°C. Do not freeze.
Polyclonal
Human, Monkey, Murine, Rat
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against recombinant CALB2.
In phosphate buffer, pH 7.3 (0.09% sodium azide)
Liquid
CALB2
Recombinant protein corresponding to human CALB2.
RUO
Primary
794
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only