Learn More
CAMK2G (phospho T286), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CAMK2G.
Manufacturer:  Abnova Corporation PAB17898
Catalog No. 89-119-540
Add to cart
Description
Description
The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a gamma chain. Six alternatively spliced variants that encode six different isoforms have been characterized to date. Additional alternative splice variants that encode different isoforms have been described, but their full-length nature has not been determined. [provided by RefSeq
Specifications
Specifications
CAMK2G
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
CAMK2G
Liquid
CAMK2G
Synthetic phosphopeptide corresponding to residues surrounding T286 of human CAMK2G.
100 uL
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
The anti-Phospho-CaMKII antibody recognizes only the phosphorylated CaMKII (Thr286) in samples from human, mouse, and rat origins.
Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CAMK2G.
In PBS (50% glycerol, 1% BSA, 0.02% thimerosal)
ACZ95102.1
CAMK|CAMK-II|CAMKG|FLJ16043|MGC26678
Rabbit
IgG
RUO
Primary
818
Human, Murine, Rat