Learn More
Cell Biolabs Inc Rabbit Anti-Carbamyl-Lysine Polyclonal Antibody, 50 µg
Supplier:  Cell Biolabs Inc STA078
Encompass_Preferred
Carbamylation is a post-translational modification which occurs throughout the lifespan of proteins in vivo. Carbamylation results from the binding of isocyanic acid, spontaneously derived from high concentrations of urea and leading to the formation of carbamyl-lysine (CBL). The carbamylation of proteins is usually associated with a partial or complete loss of protein function. It is known that elevated urea directly induces the formation of potentially atherogenic carbamylated LDL (cLDL). High blood concentrations of urea leading to the carbamylation process were detected in uremic patients and patients with end-stage renal disease. Cell Biolabs' Rabbit Anti-CBL Polyclonal Antibody is Affinity purified and suitable for Western blot or ELISA. Suitable for use with any species. 50 ug/vial.
Catalog No. NC1499597
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.