Learn More
Rabbit anti-Carbonic Anhydrase II/CA2, Biotin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP1777420.1MG
Catalog No. NBP17774201
Add to cart
Description
Description
Carbonic Anhydrase II/CA2 Polyclonal antibody specifically detects Carbonic Anhydrase II/CA2 in Human, Bovine samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Carbonic Anhydrase II/CA2
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase
Rabbit
IgG
0.1 mg
Hypoxia, Lipid and Metabolism
Primary
760
Bovine, Human
ELISA, Western Blot
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000
Purified
CA2
Full length native Carbonic Anhydrase II (purified).
Delipidation and Defibrination
RUO
Store at 4C. Do not freeze.
Polyclonal
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Carbonic Anhydrase II [Bovine Erythrocytes].