Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAT Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Chloramphenicol acetyl transferase.
Supplier: Abnova Corporation PAB10185
Description
Sequence: KTVKKNKHKFYSpecifications
CAT | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Chloramphenicol acetyl transferase. | |
Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
A synthetic peptide corresponding to amino acids 46-56 of bacteria Chloramphenicol acetyl transferase | |
RUO | |
Bacteria | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
ELISA (1:5000-1:25000) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μL | |
Primary | |
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction