Learn More
CBLN3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CBLN3.
Manufacturer: Abnova Corporation PAB17663
Description
Members of the precerebellin family, such as CBLN3, contain a cerebellin motif (see CBLN1; MIM 600432) and a C-terminal C1q signature domain (see MIM 120550) that mediates trimeric assembly of atypical collagen complexes. However, precerebellins do not contain a collagen motif, suggesting that they are not conventional components of the extracellular matrix (Pang et al., 2000 [PubMed 10964938]).[supplied by OMIM
Specifications
CBLN3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CBLN3. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q6UW01 | |
CBLN3 | |
A synthetic peptide corresponding to internal of human CBLN3. | |
RUO | |
643866 | |
Human, Mouse |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
CBLN3 | |
PRO1486 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total CBLN3 protein. | |
Liquid |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only