Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX1 Rabbit anti-Hamster, Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of CBX1.

Manufacturer:  Abnova Corporation PAB15581

Catalog No. 89-117-655


Add to cart

Description

Description

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family . The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Sequence: NEDDDKKDDKN
Specifications

Specifications

CBX1
Unconjugated
Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user.
CBX1
CBX|HP1-BETA|HP1Hs-beta|HP1Hsbeta|M31|MOD1|p25beta
Rabbit
50 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Hamster, Human
ChIP assay, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of CBX1.
In 120mM sodium phosphate buffer, pH 7.4 (50% glycerol)
Liquid
CBX1
A synthetic peptide corresponding to amino acids 176-185 of human CBX1.
RUO
Primary
10951
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only