Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC67, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CCDC67.
Supplier: Abnova Corporation PAB20522
Description
Sequence: MKQNKVPRKELPHLKEEIPFELSNLNQKLEEFRAKSREWDKQEILYQTHLISLDAQQKLLSEKCNQFQKQAQSYQTQLNGKKQCLEDSSSEIPRLICDPDPNCEINERDEFIIEKLKSAVNEIALSRNKLQDSpecifications
CCDC67 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CCDC67. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q05D60 | |
CCDC67 | |
Recombinant protein corresponding to amino acids of human CCDC67. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
CCDC67 | |
FLJ25393 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
159989 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction