Learn More
Rabbit anti-CD177, Alexa Fluor 700, Clone: 1171A, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB8186N025
Catalog No. FAB8186N025
Add to cart
This item is not returnable. View return policy
Description
Description
CD177 Monoclonal antibody specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD177
1171A
Flow Cytometry 5 uL/10^6 cells
Q8R2S8
CD177, CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1 GP, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1, PRV1NB1 glycoprotein
Rabbit
IgG
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse CD177 in direct ELISAs.
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant
CD177
Human embryonic kidney cell line HEK293-derived recombinant mouse CD177 Accession # Q8R2S8
Protein A or G purified from cell culture supernatant
RUO
Primary
57126
Mouse