Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-CD34, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP1355310.1ML

Catalog No. NBP13553101

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CD34
Unconjugated
P28906
Rabbit
0.1mL
Store at 4°C. Do not freeze.
Polyclonal
This product is specific for CD34
Immunohistochemistry, Immunohistochemistry (Paraffin)
PBS, 1% BSA buffer pH 7.6
Affinity Purified
Synthetic peptide corresponding to human CD34 protein
RUO
Primary
947
Bovine, Canine, Murine, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit