Learn More
Rabbit anti-CD7, Clone: C21-Q, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179080100UL
 View more versions of this product
Catalog No. NBP17908010
Add to cart
Description
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD7
C21-Q
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:300
P09564
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Rabbit
IgG
100 ul
Cytokine Research, Signal Transduction
Primary
924
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
CD7
Peptide derived from C-terminal region of human CD7.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human