Learn More
CD70, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyconal antibody raised against synthetic peptide of CD70.
Supplier:  Abnova Corporation PAB28356
Catalog No. 89-129-858
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin sythesis. [provided by RefSeq
Specifications
Specifications
CD70
Polyclonal
Rabbit polyconal antibody raised against synthetic peptide of CD70.
In PBS (0.09% sodium azide)
P32970
CD70
A synthetic peptide corresponding to 18 amino acids at internal region of human CD70.
50 μg
Primary
BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/ml) The optimal working dilution should be determined by the end user.
CD70
CD27L/CD27LG/TNFSF7
Rabbit
Immunoaffinity chromatography
RUO
970
Human, Monkey
Liquid
Recommendations
Recommendations