Learn More
Rabbit anti-CD8 alpha, Clone: P17-V, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 1 publication
Manufacturer:  Novus Biologicals NBP179055100UL
 View more versions of this product
Catalog No. NBP17905510
Add to cart
Description
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD8 alpha
P17-V
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100-1:200
P01732
CD8, CD8 antigen, alpha polypeptide (p32), CD8a antigen, CD8a molecule, Leu2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, RPA-T8, RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone, RPA-T8 Flow, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Rabbit
IgG
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
CD8A
Peptide derived from C-terminal region of human CD8
26 kDa
100 ul
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary
925