Learn More
CD82 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD82.
Supplier:  Abnova Corporation PAB4830
Catalog No. 89-131-598
Add to Cart
Description
Description
This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
CD82
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of CD82.
In PBS (0.09% sodium azide)
NP_002222
CD82
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CD82.
400 μL
Primary
Human, Mouse
Liquid
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user.
CD82
4F9/C33/GR15/IA4/KAI1/R2/SAR2/ST6/TSPAN27
Rabbit
Protein G purification
RUO
3732
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations