Learn More
CDC2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDC2.
Supplier:  Abnova Corporation PAB3536
Catalog No. 89-131-060
Add to Cart
Description
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
CDC2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of CDC2.
In PBS (0.09% sodium azide)
NP_001777;P06493
CDC2
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CDC2.
400 μL
Primary
Human
Liquid
Flow Cytometry, Western Blot
Unconjugated
Flow Cytometry (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
CDC2
CDC28A/CDK1/DKFZp686L20222/MGC111195
Rabbit
Ammonium sulfate precipitation
RUO
983
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions