Learn More
CDC25B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDC25B.
Supplier: Abnova Corporation PAB24841
Description
CDC25B is a member of the CDC25 family of phosphatases. CDC25B activates the cyclin dependent kinase CDC2 by removing two phosphate groups and it is required for entry into mitosis. CDC25B shuttles between the nucleus and the cytoplasm due to nuclear localization and nuclear export signals. The protein is nuclear in the M and G1 phases of the cell cycle and moves to the cytoplasm during S and G2. CDC25B has oncogenic properties, although its role in tumor formation has not been determined. Multiple transcript variants for this gene exist. [provided by RefSeq
Specifications
CDC25B | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
CDC25B | |
Rabbit | |
Antigen affinity purification | |
RUO | |
994 | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of CDC25B. | |
In 1X PBS, pH 7.2 (0.05% sodium azide) | |
CDC25B | |
A synthetic peptide corresponding to CDC25B. | |
100 μL | |
Primary | |
CDC25B polyclonal antibody detects endogenous levels of CDC25B protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only