Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDKN1B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of CDKN1B.

Manufacturer:  Abnova Corporation PAB18369

Catalog No. 89-119-935


Add to cart

Description

Description

This gene encodes a cyclin-dependent kinase inhibitor, which shares a limited similarity with CDK inhibitor CDKN1A/p21. The encoded protein binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. [provided by RefSeq

Specifications

Specifications

CDKN1B
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of CDKN1B.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
P46527
CDKN4|KIP1|MEN1B|MEN4|P27KIP1
Rabbit
Affinity purification
RUO
Primary
1027
Human
ELISA, Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user.
CDKN1B
Liquid
CDKN1B
A synthetic peptide corresponding to human CDKN1B.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is specific to CDKN1B.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only