Learn More
CDKN2A Rabbit anti-Human, DyLight 488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.
Manufacturer:  Abnova Corporation PAB15354
Catalog No. 89-117-584
Add to cart
Description
Description
This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, MDM1, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene. [provided by RefSeq
Specifications
Specifications
CDKN2A
DyLight 488
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
CDKN2A
Liquid
CDKN2A
A synthetic peptide corresponding to amino acids 100-200 of human CDKN2A.
RUO
Primary
1029
Human
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.
In 50mM sodium borate
Q8N726
ARF|CDK4I|CDKN2|CMM2|INK4|INK4a|MLM|MTS1|TP16|p14|p14ARF|p16|p16INK4|p16INK4a|p19
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody reacts with human p14ARF. Other species have not been tested. This is specific to p14ARF. This antibody may be used for Western blotting, where a band is seen at ~16 KDa, representing p14ARF (p14ARF tends to run slightly higher than the theoretical MW of 14 KDa). Additional non-specific bands may be seen at ~25 and 47 KDa.