Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdkn2a, Rabbit, HRP, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.

Manufacturer:  Abnova Corporation PAB15343

Catalog No. 89-117-573


Add to cart

Description

Description

Specifications

Specifications

Cdkn2a
HRP
Western Blot (2 ug/mL) Immunocytochemistry (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user.
Cdkn2a
Liquid
Cdkn2a
A synthetic peptide (conjugated with KLH) corresponding to amino acids 50-150 of mouse Cdkn2a.
RUO
Primary
12578
Mouse
ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.
In PBS
Q64364;P51480
ARF-INK4a|Arf|INK4a-ARF|Ink4a/Arf|MTS1|Pctr1|p16|p16(INK4a)|p16INK4a|p19|p19ARF
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Cross-reacts with mouse. Not yet tested in other species. The immunogen used for this production shares 80% homology with human protein, 93% with rat protein, and 86% with porcine protein. On Western blot the detects a band corresponding to p19ARF in mouse embryonic fibroblasts. Detects a band of approximately 19 KDa.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only