Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM6, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant CEACAM6.

Manufacturer:  Abnova Corporation PAB16183

Catalog No. 89-118-073


Add to cart

Description

Description

Carcinoembryonic antigen (CEA; MIM 114890) is one of the most widely used tumor markers in serum immunoassay determinations of carcinoma. An apparent lack of absolute cancer specificity for CEA probably results in part from the presence in normal and neoplastic tissues of antigens that share antigenic determinants with the 180kDa form of CEA (Barnett et al., 1988 [PubMed 3220478]). For background information on the CEA family of genes, see CEACAM1 (MIM 109770).[supplied by OMIM

Specifications

Specifications

CEACAM6
Unconjugated
ELISA (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
CEACAM6
CD66c|CEAL|NCA
Rabbit
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human
ELISA
Rabbit polyclonal antibody raised against recombinant CEACAM6.
Lyophilized from PBS.
Lyophilized
CEACAM6
Recombinant protein corresponding to human CEACAM6.
RUO
Primary
4680
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only