Learn More
CHMP4B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CHMP4B.
Supplier: Abnova Corporation PAB27902
Description
CHMP4B belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A (MIM 164010), is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 [PubMed 16730941]).[supplied by OMIM
Sequence: SVFGKLFGAGGGKAGKGGPTPQEAIQRLRDTEEMLSKKSpecifications
CHMP4B | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CHMP4B. | |
In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
C20orf178/CHMP4A/CTPP3/SNF7/SNF7-2/Shax1/dJ553F4.4 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
128866 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry, Western Blot, Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
CHMP4B | |
CHMP4B | |
Recombinant protein corresponding to amino acids of human CHMP4B. | |
100 μL | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only