Learn More
CHRM3 Rabbit anti-Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHRM3.
Manufacturer:  Abnova Corporation PAB16212
Catalog No. 89-118-102
Add to cart
Description
Description
The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue. [provided by RefSeq
Specifications
Specifications
CHRM3
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
CHRM3
Liquid
CHRM3
A synthetic peptide (conjugated with KLH) corresponding to human CHRM3.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
3rd cytoplasmic domain of human. Predicted crossreactivity with mouse due to sequence similarity .
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of CHRM3.
In PBS (0.09% sodium azide)
P20309
HM3
Rabbit
Immunoaffinity purification
RUO
Primary
1131
Bovine, Canine, Equine, Hamster, Human, Monkey, Murine, Porcine, Rabbit, Rat, Sheep