Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHUK Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of CHUK.

Manufacturer:  Abnova Corporation PAB13399

Catalog No. 89-116-645


Add to cart

Description

Description

This gene encodes a member of the serine/threonine protein kinase family. The encoded protein, a component of a cytokine-activated protein complex that is an inhibitor of the essential transcription factor NF-kappa-B complex, phosphorylates sites that trigger the degradation of the inhibitor via the ubiquination pathway, thereby activating the transcription factor. [provided by RefSeq

Specifications

Specifications

CHUK
Unconjugated
Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
CHUK
Liquid
CHUK
A synthetic peptide corresponding to amino acids 716-734 of human CHUK.
RUO
Primary
1147
Human
Immunocytochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of CHUK.
In PBS (0.02% sodium azide)
AF009225
IKBKA|IKK-alpha|IKK1|IKKA|NFKBIKA|TCF16
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
human IKK alpha differs from corresponding murine sequence by one amino acid. An 85 KDa band should be detected. This antibody has no cross response to IKKb or IKKg.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only