Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIC Rabbit anti-Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant CIC.

Manufacturer:  Abnova Corporation PAB12458

Catalog No. 89-115-832


Add to cart

Description

Description

Specifications

Specifications

CIC
Unconjugated
Immunoprecipitation (2-5 ug/mg lysate) Western Blot (1:1000-1:2500) The optimal working dilution should be determined by the end user.
CIC
Liquid
CIC
Recombinant protein corresponding to amino acids 1500-1608 of human CIC.
RUO
Primary
23152
Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against partial recombinant CIC.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Q96RK0
KIAA0306
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Canine, Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only