Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cideb, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Cideb.

Manufacturer:  Abnova Corporation PAB11961

Catalog No. 89-115-474


Add to cart

Description

Description

Specifications

Specifications

Cideb
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Cideb
Liquid
Cideb
A synthetic peptide corresponding to amino acids 204-219 of mouse Cideb.
RUO
Primary
12684
Mouse
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Cideb.
In PBS, pH 7.4 (0.02% sodium azide)
AF041377
1110030C18Rik|AI790179|CIDE-B
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes mouse CIDE-B (25kD). No cross-reactivity with CIDE-A. Epitopes at the C-terminus of CIDE-B.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only