Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Claudin-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP1356520.1ML

Catalog No. NBP13565201


Add to cart

Description

Description

Claudin-1 Polyclonal antibody specifically detects Claudin-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Claudin-1
Unconjugated
PBS (pH 7.6) and 1.0% BSA with Sodium Azide
Affinity Purified
CLDN1
Synthetic peptide derived from C-terminus of human Claudin-1 peptide.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Claudin 1
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200
O95832
claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
Rabbit
IgG
0.1 ml
Extracellular Matrix
Primary
9076
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit