Learn More
Col2a1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Col2a1.
Manufacturer:  Abnova Corporation PAB19147
Catalog No. 89-120-676
Add to cart
Description
Description
Specifications
Specifications
Col2a1
Unconjugated
Western blot (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Col2a1
Col2|Col2a|Col2a-1|Del1|Dmm|MGC90638
Rabbit
Affinity purification
RUO
Primary
12824
Human, Murine, Rat
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against native Col2a1.
In PBS
Liquid
Col2a1
Native purified Col2a1 CnBr fragment from mouse chondrosarcoma.
100 ug
Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
It exhibits very moderately with bovine collagen type XI and slightly with collagen type I (< 10%).