Learn More
COL4A1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COL4A1.
Supplier: Abnova Corporation PAB17326
Description
This gene encodes the major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter. [provided by RefSeq
Specifications
COL4A1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of COL4A1. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
P02462 | |
COL4A1 | |
A synthetic peptide corresponding to amino acids 31-70 of human COL4A1. | |
RUO | |
1282 | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:300-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
COL4A1 | |
arresten | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total COL4A1 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only