Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL9A1/COL9A2/COL9A3, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against native COL9A1/COL9A2/COL9A3.

Manufacturer:  Abnova Corporation PAB13511

Catalog No. 89-116-714


Add to cart

Description

Description

Specifications

Specifications

COL9A1/COL9A2/COL9A3
Unconjugated
Immunofluorescence (1:40) The optimal working dilution should be determined by the end user.
Lyophilized
Rabbit
500 uL
Store at 4°C on dry atmosphere. After reconstitution with 0.5mL distilled water
Polyclonal
This antibody reacts with chicken collagen type IX 100%, chicken collagen type I, II, III and V negative in ELISA at 1:100 dilution.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays
Rabbit polyclonal antibody raised against native COL9A1/COL9A2/COL9A3.
Lyophilized from purified antibody
COL9A1
Native purified COL9A1/COL9A2/COL9A3 from chicken breast bone.
RUO
Primary
771873|396524|396242
Chicken
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only