Learn More
Collagen Type I Rabbit anti-Fish, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Collagen Type I.
Manufacturer:  Abnova Corporation PAB13491
Catalog No. 89-137-607
Add to cart
Description
Description
Specifications
Specifications
Collagen Type I
Unconjugated
Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user.
Lyophilized
Native purified Collagen Type I from tuna fish skin.
RUO
Primary
This antibody reacts with tuna fish collagen type I.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays
Rabbit polyclonal antibody raised against native Collagen Type I.
Lyophilized from phosphate buffer
Rabbit
100 uL
Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Fish