Learn More
Collagen Type IV Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Collagen Type IV.
Manufacturer:  Abnova Corporation PAB13505
Catalog No. 89-137-611
Add to cart
Description
Description
Specifications
Specifications
Collagen Type IV
Unconjugated
Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user.
Lyophilized
Native purified Collagen Type IV from mouse tumour tissue.
RUO
Primary
This antibody reacts with murine collagen type IV 100%; murine collagen type I, II and III <0.1%; human collagen type IV and V <0.1%; murine fibronectin <0.1%; murine laminin <0.1% in RIA at 1:5000 dilution.
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays
Rabbit polyclonal antibody raised against native Collagen Type IV.
Lyophilized from PBS
Rabbit
100 uL
Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Polyclonal
Mouse