Learn More
Rabbit anti-Complement C3d, Clone: E28-P, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179074100UL
 View more versions of this product
Catalog No. NBP17907410
Add to cart
Description
Description
Complement C3d Monoclonal antibody specifically detects Complement C3d in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Complement C3d
E28-P
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200
P01024
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Rabbit
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
C3
Peptide derived from N-terminal sequence of human C3.
100 ul
Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience
Primary
718