Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CORT Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of CORT.

Supplier:  Abnova Corporation PAB4980

Catalog No. 89-131-663


Add to Cart

Description

Description

The product of this gene is a neuropeptide with strong structural similarity to somatostatin. It binds to all known somatostatin receptors, and shares many pharmacological and functional properties with somatostatin, including the depression of neuronal activity. However, it also has many properties distinct from somatostatin, such as induction of slow-wave sleep, apparently by antagonism of the excitatory effects of acetylcholine on the cortex, reduction of locomotor activity, and activation of cation selective currents not responsive to somatostatin. [provided by RefSeq

Sequence: PCKNFFWKTFSSCK
Specifications

Specifications

CORT
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of CORT.
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
CST-14/CST-17/CST-29
Rabbit
150 μg
Primary
Human
Liquid
ELISA, Immunoprecipitation
Unconjugated
The optimal working dilution should be determined by the end user.
CORT
CORT
A synthetic peptide (conjugated with KLH) corresponding to human CORT.
RUO
1325
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only