Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Corticotropin Releasing Factor, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP1357030.1ML

Catalog No. NBP13570301

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Corticotropin Releasing Factor
0.2 mg/mL
Western Blot, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100
Affinity Purified
CRH
A synthetic peptide from C-terminus of human CRF protein
4.5 kDa
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Corticotropin Releasing Factor
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.6) and 1.0% BSA with 0.1% Sodium Azide
corticoliberin, corticotropin releasing hormone, Corticotropin-releasing hormone, CRFCorticotropin-releasing factor
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
1392
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit