Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-COX-2, Clone: SP21, Novus Biologicals™

Rabbit Monoclonal Antibody

Specifications

Antigen COX-2
Clone SP21
Immunogen Synthetic peptide corresponding to C-terminus of rat COX2.
Gene ID (Entrez) 5743
Formulation TBS (pH 7.5) and 1.0% BSA with 0.1% Sodium Azide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14979601
View Documents
Novus Biologicals
NBP1497960.1ML
0.1 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

COX-2 Monoclonal antibody specifically detects COX-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

COX-2
Synthetic peptide corresponding to C-terminus of rat COX2.
TBS (pH 7.5) and 1.0% BSA with 0.1% Sodium Azide
Primary
Rabbit
Unconjugated
Tissue culture supernatant
Immunohistochemistry, Immunohistochemistry (Paraffin)
SP21
5743
Monoclonal
Store at 4C. Do not freeze.
IgG
Supernatant
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit