Learn More
CPT1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CPT1B.
Supplier: Abnova Corporation PAB4375
Description
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene. [provided by RefSeq
Specifications
CPT1B | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CPT1B. | |
In PBS (0.09% sodium azide) | |
CPTM_HUMAN | |
CPT1B | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CPT1B. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:50-200) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
CPT1B | |
CPT1-M/KIAA1670/M-CPT1/MCCPT1 | |
Rabbit | |
Protein G purification | |
RUO | |
1375 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only