Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREB3L1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CREB3L1.
Supplier: Abnova Corporation PAB4813
Specifications
CREB3L1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CREB3L1. | |
In PBS (0.09% sodium azide) | |
NP_443086 | |
CREB3L1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CREB3L1. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
CREB3L1 | |
OASIS | |
Rabbit | |
Protein G purification | |
RUO | |
90993 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only