Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crlf2, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Crlf2.

Manufacturer:  Abnova Corporation PAB13268

Catalog No. 89-116-516


Add to cart

Description

Description

Specifications

Specifications

Crlf2
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Crlf2
Liquid
Crlf2
A synthetic peptide corresponding to internal region 21 amino acids of mouse Crlf2.
RUO
Primary
57914
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Crlf2.
In PBS (0.02% sodium azide)
NP_057924
CRLM2|Ly114|Tpte2|Tslpr
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only