Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSD Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant CTSD.

Manufacturer:  Abnova Corporation PAB12789

Catalog No. 89-116-092


Add to cart

Description

Description

This gene encodes a lysosomal aspartyl protease composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. This proteinase, which is a member of the peptidase C1 family, has a specificity similar to but narrower than that of pepsin A. Transcription of this gene is initiated from several sites, including one which is a start site for an estrogen-regulated transcript. Mutations in this gene are involved in the pathogenesis of several diseases, including breast cancer and possibly Alzheimer disease. [provided by RefSeq

Specifications

Specifications

CTSD
1 mg/mL
Rabbit polyclonal antibody raised against recombinant CTSD.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
CTSD
Recombinant His fusion protein corresponding to human CTSD.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Rat
Western Blot
Unconjugated
Western Blot (1:5000) The optimal working dilution should be determined by the end user.
CTSD
CLN10|CPSD|MGC2311
Rabbit
Affinity purification
RUO
Primary
1509
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only